accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 5655-61-8

IUPAC Name: (1S,2R,4S)-1,7,7-trimethylbicyclo[2.2.1]heptan-2-yl acetate
Synonyms: L-bornyl acetate
Molecular Weight (g/mol): 196.29
Molecular Formula: C12H20O2
InChi Key: KGEKLUUHTZCSIP-HOSYDEDBSA-N
SMILES: CC(=O)O[C@@H]1C[C@@H]2CC[C@@]1(C)C2(C)C
Molecular Weight (g/mol) 
  • (8)
  • (2)
Quantity 
  • (3)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
Boiling Point 
  • (9)
Percent Purity 
  • (1)
  • (7)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (7)
  • (2)
  • (1)
  • (7)
  • (8)
  • (2)
  • (3)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (9)
  • (1)
  • (7)
  • (5)