accessibility menu, dialog, popup

UserName

Viewing profile as user:
ClClClClCl

CAS RN 56558-18-0

IUPAC Name: 2,3',4,5',6-pentachloro-1,1'-biphenyl
Synonyms: 2,3',4,5',6-pentachloro-1,1'-biphenyl
Molecular Weight (g/mol): 326.42
Molecular Formula: C12H5Cl5
InChi Key: XBVSGJGMWSKAKL-UHFFFAOYSA-N
SMILES: ClC1=CC(Cl)=C(C(Cl)=C1)C1=CC(Cl)=CC(Cl)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results