accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3C(S)OOCH3NH2HCl

CAS RN 56558-30-6

IUPAC Name: methyl (2S)-2-aminopentanoate hydrochloride
Synonyms: methyl (2S)-2-aminopentanoate hydrochloride| H-NVA-OME HCL
Molecular Weight (g/mol): 167.63
Molecular Formula: C6H14ClNO2
InChi Key: IODNYRVZHKRFIW-JEDNCBNOSA-N
SMILES: Cl.CCC[C@H](N)C(=O)OC

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)