accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 5660-45-7

SSSSS

CAS RN 5660-45-7

IUPAC Name: 5-{[2,2'-bithiophen]-5-yl}-5'-(thiophen-2-yl)-2,2'-bithiophene
Synonyms: 5-{[2,2'-bithiophen]-5-yl}-5'-(thiophen-2-yl)-2,2'-bithiophene
Molecular Weight (g/mol): 412.62
Molecular Formula: C20H12S5
InChi Key: YFBLUJZFRBFQMR-UHFFFAOYSA-N
SMILES: S1C=CC=C1C1=CC=C(S1)C1=CC=C(S1)C1=CC=C(S1)C1=CC=CS1
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results