CAS RN 5660-91-3
IUPAC Name:
1,3-diphenyl-2H-cyclopenta[l]phenanthren-2-one
Synonyms:
1,3-diphenylcyclopenta[l]phenanthren-2-one
Molecular Weight (g/mol):
382.46
Molecular Formula:
C29H18O
InChi Key:
MNSDGJFEKUKHGO-UHFFFAOYSA-N
SMILES:
O=C1C(=C2C(=C1C1=CC=CC=C1)C1=CC=CC=C1C1=CC=CC=C21)C1=CC=CC=C1
1
–
1
of
1
results
1
Promotions Available
Phencyclone, 97%
Phencyclone, 97%
CAS: 5660-91-3 Molecular Formula: C29H18O Molecular Weight (g/mol): 382.46 MDL Number: MFCD00051451 InChI Key: MNSDGJFEKUKHGO-UHFFFAOYSA-N Synonym: phencyclone,1,3-diphenyl-2h-cyclopenta l phenanthren-2-one,1,3-diphenylcyclopenta l phenanthren-2-one,1,3-diphenylcyclopenta 1,2-l phenanthren-2-one PubChem CID: 97140 ChEBI: CHEBI:48304 IUPAC Name: 1,3-diphenylcyclopenta[l]phenanthren-2-one SMILES: O=C1C(=C2C(=C1C1=CC=CC=C1)C1=CC=CC=C1C1=CC=CC=C21)C1=CC=CC=C1