accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 56601-42-4

IUPAC Name: isothiocyanatocyclopropane
Synonyms: isothiocyanatocyclopropane
Molecular Weight (g/mol): 99.15
Molecular Formula: C4H5NS
InChi Key: JGFBQFKZKSSODQ-UHFFFAOYSA-N
SMILES: S=C=NC1CC1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results