accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 56613-81-1

IUPAC Name: (1S)-2-amino-1-phenylethan-1-ol
Synonyms: (1S)-2-amino-1-phenylethanol
Molecular Weight (g/mol): 137.18
Molecular Formula: C8H11NO
InChi Key: ULSIYEODSMZIPX-MRVPVSSYSA-N
SMILES: NC[C@@H](O)C1=CC=CC=C1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results