accessibility menu, dialog, popup

UserName

Viewing profile as user:
NNNN

CAS RN 56643-83-5

IUPAC Name: 1-({4-[(1H-imidazol-1-yl)methyl]phenyl}methyl)-1H-imidazole
Synonyms: 1-{[4-(imidazol-1-ylmethyl)phenyl]methyl}imidazole
Molecular Weight (g/mol): 238.29
Molecular Formula: C14H14N4
InChi Key: NKUFFYFOBGGDTP-UHFFFAOYSA-N
SMILES: C(N1C=CN=C1)C1=CC=C(CN2C=CN=C2)C=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results