accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CNCH3H3CH3CH3CNCH3H3CH3COCrOOO

CAS RN 56660-19-6

IUPAC Name: dioxochromiumbis(olate); bis(tetrabutylazanium)
Synonyms: chromate(2-); bis(tetrabutylammonium ion)
Molecular Weight (g/mol): 600.93
Molecular Formula: C32H72CrN2O4
InChi Key: ZWQUOQAMKSRLDH-UHFFFAOYSA-N
SMILES: [O-][Cr]([O-])(=O)=O.CCCC[N+](CCCC)(CCCC)CCCC.CCCC[N+](CCCC)(CCCC)CCCC
Molecular Weight (g/mol) 
  • (1)
  • (1)
Quantity 
  • (2)
Percent Purity 
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)