accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 5668-93-9

CH3

CAS RN 5668-93-9

IUPAC Name: 3-ethyl-1,1'-biphenyl
Synonyms: 3-ethyl-1,1'-biphenyl
Molecular Weight (g/mol): 182.27
Molecular Formula: C14H14
InChi Key: HUXKTWJQSHBZIV-UHFFFAOYSA-N
SMILES: CCC1=CC=CC(=C1)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results