accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 5669-19-2

IUPAC Name: 2-benzylprop-2-enoic acid
Synonyms: 2-benzylprop-2-enoic acid
Molecular Weight (g/mol): 162.19
Molecular Formula: C10H10O2
InChi Key: RYNDYESLUKWOEE-UHFFFAOYSA-N
SMILES: OC(=O)C(=C)CC1=CC=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results