accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 566939-58-0

IUPAC Name: 1,3-dibromo-5-octyl-4H,5H,6H-thieno[3,4-c]pyrrole-4,6-dione
Synonyms: 1,3-dibromo-5-octylthieno[3,4-c]pyrrole-4,6-dione
Molecular Weight (g/mol): 423.16
Molecular Formula: C14H17Br2NO2S
InChi Key: GSGMEQUXTCYOAU-UHFFFAOYSA-N
SMILES: CCCCCCCCN1C(=O)C2=C(Br)SC(Br)=C2C1=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results