accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3HOHOO

CAS RN 567-61-3

IUPAC Name: 2-hydroxy-6-methylbenzoic acid
Synonyms: methylsalicylic acid 6-msa
Molecular Weight (g/mol): 152.15
Molecular Formula: C8H8O3
InChi Key: HCJMNOSIAGSZBM-UHFFFAOYSA-N
SMILES: CC1=CC=CC(O)=C1C(O)=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results