accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 5672-83-3

IUPAC Name: (4S)-4-{[(benzyloxy)carbonyl]amino}-5-methoxy-5-oxopentanoate
Synonyms: (4S)-4-{[(benzyloxy)carbonyl]amino}-5-methoxy-5-oxopentanoate
Molecular Weight (g/mol): 294.28
Molecular Formula: C14H16NO6
InChi Key: BGMCTGARFXPQML-NSHDSACASA-M
SMILES: COC(=O)[C@H](CCC([O-])=O)NC(=O)OCC1=CC=CC=C1
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)