accessibility menu, dialog, popup

UserName

Viewing profile as user:
OFFF

CAS RN 56734-74-8

IUPAC Name: 2-(trifluoromethyl)cyclohexan-1-one
Synonyms: 2-(trifluoromethyl)cyclohexan-1-one
Molecular Weight (g/mol): 166.14
Molecular Formula: C7H9F3O
InChi Key: RZSQHJQIBOWYSQ-UHFFFAOYNA-N
SMILES: FC(F)(F)C1CCCCC1=O
Quantity 
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)