accessibility menu, dialog, popup

UserName

Viewing profile as user:
FN(E/Z)

CAS RN 5676-81-3

IUPAC Name: 1-(4-fluorophenyl)-N-phenylmethanimine
Synonyms: 1-(4-fluorophenyl)-N-phenylmethanimine
Molecular Weight (g/mol): 199.23
Molecular Formula: C13H10FN
InChi Key: MPRONVWLCPZXOB-UHFFFAOYSA-N
SMILES: FC1=CC=C(C=NC2=CC=CC=C2)C=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results