accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 56794-28-6

IUPAC Name: 4-chloro-N,N-diethylbutanamide
Synonyms: 4-chloro-N,N-diethylbutanamide
Molecular Weight (g/mol): 177.67
Molecular Formula: C8H16ClNO
InChi Key: HHBZYNJHOOGYPZ-UHFFFAOYSA-N
SMILES: CCN(CC)C(=O)CCCCl

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results