accessibility menu, dialog, popup

UserName

Viewing profile as user:
CeH3CCH3OOH3CCH3OOH3CCH3OO

CAS RN 56797-01-4

IUPAC Name: cerium(3+) tris(2-ethylhexanoate)
Synonyms: cerium(3+) tris(2-ethylhexanoate)
Molecular Weight (g/mol): 569.73
Molecular Formula: C24H45CeO6
InChi Key: GGVUYAXGAOIFIC-UHFFFAOYNA-K
SMILES: [Ce+3].CCCCC(CC)C([O-])=O.CCCCC(CC)C([O-])=O.CCCCC(CC)C([O-])=O
Physical Form 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (1)
  • (1)
  • (1)