accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3SiCH3CH3OHO

CAS RN 5683-31-8

IUPAC Name: 3-(trimethylsilyl)prop-2-ynoic acid
Synonyms: 3-(trimethylsilyl)prop-2-ynoic acid
Molecular Weight (g/mol): 142.23
Molecular Formula: C6H10O2Si
InChi Key: IPEATTYBFBRNEB-UHFFFAOYSA-N
SMILES: C[Si](C)(C)C#CC(O)=O
Percent Purity 
  • (2)
  • (3)
  • (2)
Quantity 
  • (3)
  • (1)
  • (3)
Boiling Point 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (2)
  • (3)
  • (2)
  • (3)
  • (2)
  • (3)
  • (1)
  • (3)
  • (2)
  • (2)
  • (2)