accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 568577-87-7

IUPAC Name: 1,3-dimethyl-5-(morpholin-4-yl)-1H-pyrazol-4-amine
Synonyms: 1,3-dimethyl-5-(morpholin-4-yl)pyrazol-4-amine
Molecular Weight (g/mol): 196.25
Molecular Formula: C9H16N4O
InChi Key: LEVZXNJICHTQEC-UHFFFAOYSA-N
SMILES: CN1N=C(C)C(N)=C1N1CCOCC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results