accessibility menu, dialog, popup

UserName

Viewing profile as user:
OO

CAS RN 5690-03-9

IUPAC Name: 1H,2H,3H-naphtho[2,1-b]pyran-3-one
Synonyms: 1H,2H-naphtho[2,1-b]pyran-3-one
Molecular Weight (g/mol): 198.22
Molecular Formula: C13H10O2
InChi Key: ISFPDBUKMJDAJH-UHFFFAOYSA-N
SMILES: O=C1CCC2=C3C=CC=CC3=CC=C2O1
Color 
  • (2)
Grade 
  • (2)
Percent Purity 
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results