accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 56908-88-4

IUPAC Name: 1,3-dibromo-5-(bromomethyl)benzene
Synonyms: 1,3-dibromo-5-(bromomethyl)benzene
Molecular Weight (g/mol): 328.83
Molecular Formula: C7H5Br3
InChi Key: PWTFRUXTAFBWBW-UHFFFAOYSA-N
SMILES: BrCC1=CC(Br)=CC(Br)=C1
Percent Purity 
  • (2)
  • (4)
Quantity 
  • (1)
  • (2)
  • (3)
Physical Form 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (2)
  • (4)
  • (2)
  • (4)
  • (1)
  • (2)
  • (3)
  • (2)
  • (1)
  • (2)