accessibility menu, dialog, popup

UserName

Viewing profile as user:
NaNaOSOOHNNNOSOOO(E)

CAS RN 56932-43-5

IUPAC Name: disodium (7E)-8-oxo-7-[2-(6-sulfonatonaphthalen-2-yl)hydrazin-1-ylidene]-7,8-dihydroquinoline-5-sulfonate
Synonyms: disodium (7E)-8-oxo-7-[2-(6-sulfonatonaphthalen-2-yl)hydrazin-1-ylidene]quinoline-5-sulfonate
Molecular Weight (g/mol): 503.41
Molecular Formula: C19H11N3Na2O7S2
InChi Key: YOGRUDWAJPVHEL-LLDDCTHSSA-L
SMILES: [Na+].[Na+].[O-]S(=O)(=O)C1=CC=C2C=C(N\N=C3/C=C(C4=CC=CN=C4C3=O)S([O-])(=O)=O)C=CC2=C1
Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results