accessibility menu, dialog, popup

UserName

Viewing profile as user:
SSOOClClCl

CAS RN 56946-83-9

IUPAC Name: 2,5-dichlorothiophene-3-sulfonyl chloride
Synonyms: 2,5-dichlorothiophene-3-sulfonyl chloride
Molecular Weight (g/mol): 251.52
Molecular Formula: C4HCl3O2S2
InChi Key: JJKSHSHZJOWSEC-UHFFFAOYSA-N
SMILES: ClC1=CC(=C(Cl)S1)S(Cl)(=O)=O
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results