CAS RN 5696-15-1
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Butoxamine hydrochloride, MilliporeSigma™ Supelco™
Butoxamine hydrochloride is a β-andrenoceptor antagonist, which shows β2-selectivity.