accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 56961-77-4

IUPAC Name: 1-bromo-2,3-dichlorobenzene
Synonyms: 1-bromo-2,3-dichlorobenzene
Molecular Weight (g/mol): 225.89
Molecular Formula: C6H3BrCl2
InChi Key: HVKCZUVMQPUWSX-UHFFFAOYSA-N
SMILES: ClC1=CC=CC(Br)=C1Cl
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (2)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (4)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)