accessibility menu, dialog, popup

UserName

Viewing profile as user:
OHClO

CAS RN 56962-11-9

IUPAC Name: 2-chloro-4-hydroxybenzaldehyde
Synonyms: 2-chloro-4-hydroxybenzaldehyde
Molecular Weight (g/mol): 156.57
Molecular Formula: C7H5ClO2
InChi Key: ZMOMCILMBYEGLD-UHFFFAOYSA-N
SMILES: OC1=CC(Cl)=C(C=O)C=C1
Molecular Weight (g/mol) 
  • (2)
  • (1)
Quantity 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results