CAS RN 56970-78-6
CAS RN 56970-78-6
IUPAC Name:
3-bromo-2-methylpropanoic acid
Synonyms:
3-bromo-2-methylpropanoic acid
Molecular Weight (g/mol):
167
Molecular Formula:
C4H7BrO2
InChi Key:
BUPXDXGYFXDDAA-UHFFFAOYNA-N
SMILES:
CC(CBr)C(O)=O
1
–
1
of
1
results
1
3-Bromoisobutyric Acid 97.0+%, TCI America™
CAS: 56970-78-6 Molecular Formula: C4H7BrO2 Molecular Weight (g/mol): 167.00 MDL Number: MFCD01310530 InChI Key: BUPXDXGYFXDDAA-UHFFFAOYNA-N Synonym: 3-Bromo-2-methylpropionic Acid PubChem CID: 304776 IUPAC Name: 3-bromo-2-methylpropanoic acid SMILES: CC(CBr)C(O)=O