accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 5699-40-1

IUPAC Name: N-(diaminomethylidene)acetamide
Synonyms: N-(diaminomethylidene)acetamide
Molecular Weight (g/mol): 101.11
Molecular Formula: C3H7N3O
InChi Key: NGGXACLSAZXJGM-UHFFFAOYSA-N
SMILES: CC(=O)N=C(N)N

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results