accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH2NCS

CAS RN 57-06-7

IUPAC Name: 3-isothiocyanatoprop-1-ene
Synonyms: allyl isothiocyanate caswell no. 027 senfoel mustard oil, volatile fema number 2034 carbospol oleum sinapis volatile artificial oil of mustard volatile mustard oil allyspol
Molecular Weight (g/mol): 99.15
Molecular Formula: C4H5NS
InChi Key: ZOJBYZNEUISWFT-UHFFFAOYSA-N
SMILES: C=CCN=C=S
Boiling Point 
  • (3)
  • (3)
  • (3)
Molecular Weight (g/mol) 
  • (5)
  • (4)
Percent Purity 
  • (6)
  • (3)
Quantity 
  • (2)
  • (1)
  • (3)
  • (1)
  • (2)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (3)
  • (3)
  • (3)
  • (3)
  • (5)
  • (4)
  • (6)
  • (3)
  • (2)
  • (1)
  • (3)
  • (1)
  • (2)
  • (3)