CAS RN 57-13-6
Urea (Colorless-to-White Crystals or Crystalline Powder/Mol. Biol.), Fisher BioReagents™
CAS: 57-13-6 Molecular Formula: CH4N2O Molecular Weight (g/mol): 60.056 InChI Key: XSQUKJJJFZCRTK-UHFFFAOYSA-N Synonym: carbamide,isourea,carbonyldiamide,ureophil,carbonyldiamine,carbamimidic acid,pseudourea,ureaphil,urevert,alphadrate PubChem CID: 1176 ChEBI: CHEBI:48376 IUPAC Name: urea SMILES: C(=O)(N)N
Urea, 98+%
CAS: 57-13-6 Molecular Formula: CH4N2O Molecular Weight (g/mol): 60.056 MDL Number: MFCD00008022 InChI Key: XSQUKJJJFZCRTK-UHFFFAOYSA-N Synonym: carbamide,isourea,carbonyldiamide,ureophil,carbonyldiamine,carbamimidic acid,pseudourea,ureaphil,urevert,alphadrate PubChem CID: 1176 ChEBI: CHEBI:48376 IUPAC Name: urea SMILES: C(=O)(N)N
Urea, ultrapure, 99%
CAS: 57-13-6 Molecular Formula: CH4N2O Molecular Weight (g/mol): 60.056 MDL Number: MFCD00008022 InChI Key: XSQUKJJJFZCRTK-UHFFFAOYSA-N Synonym: carbamide,isourea,carbonyldiamide,ureophil,carbonyldiamine,carbamimidic acid,pseudourea,ureaphil,urevert,alphadrate PubChem CID: 1176 ChEBI: CHEBI:48376 IUPAC Name: urea SMILES: C(=O)(N)N
Urea, 99%, ACS reagent
CAS: 57-13-6 MDL Number: MFCD00008022 InChI Key: XSQUKJJJFZCRTK-UHFFFAOYSA-N Synonym: carbamide,isourea,carbonyldiamide,ureophil,carbonyldiamine,carbamimidic acid,pseudourea,ureaphil,urevert,alphadrate PubChem CID: 1176 ChEBI: CHEBI:48376 IUPAC Name: urea SMILES: C(=O)(N)N
Thermo Scientific Chemicals Urea, 99.5%, for molecular biology, DNAse, RNAse and Protease free
CAS: 57-13-6 Molecular Formula: CH4N2O Molecular Weight (g/mol): 60.06 MDL Number: MFCD00008022 InChI Key: XSQUKJJJFZCRTK-UHFFFAOYSA-N Synonym: carbamide,isourea,carbonyldiamide,ureophil,carbonyldiamine,carbamimidic acid,pseudourea,ureaphil,urevert,alphadrate PubChem CID: 1176 ChEBI: CHEBI:48376 IUPAC Name: urea SMILES: C(=O)(N)N
Urea (USP), Fisher Chemical™
CAS: 57-13-6 Molecular Formula: CH4N2O Molecular Weight (g/mol): 60.056 MDL Number: MFCD00008022 InChI Key: XSQUKJJJFZCRTK-UHFFFAOYSA-N Synonym: carbamide,isourea,carbonyldiamide,ureophil,carbonyldiamine,carbamimidic acid,pseudourea,ureaphil,urevert,alphadrate PubChem CID: 1176 ChEBI: CHEBI:48376 IUPAC Name: urea SMILES: C(=O)(N)N
Urea (Certified ACS), Fisher Chemical™
CAS: 57-13-6 Molecular Formula: CH4N2O Molecular Weight (g/mol): 60.056 MDL Number: MFCD00008022 InChI Key: XSQUKJJJFZCRTK-UHFFFAOYSA-N Synonym: carbamide,isourea,carbonyldiamide,ureophil,carbonyldiamine,carbamimidic acid,pseudourea,ureaphil,urevert,alphadrate PubChem CID: 1176 ChEBI: CHEBI:48376 IUPAC Name: urea SMILES: C(=O)(N)N
Urea, ≥99.5%, Molecular Biology Grade, Ultrapure
CAS: 57-13-6 Molecular Formula: CH4N2O Molecular Weight (g/mol): 60.056 MDL Number: MFCD00008022 InChI Key: XSQUKJJJFZCRTK-UHFFFAOYSA-N Synonym: carbamide,isourea,carbonyldiamide,ureophil,carbonyldiamine,carbamimidic acid,pseudourea,ureaphil,urevert,alphadrate PubChem CID: 1176 ChEBI: CHEBI:48376 IUPAC Name: urea SMILES: C(=O)(N)N
Urea, ACS, 99.0-100.5%
CAS: 57-13-6 Molecular Formula: CH4N2O Molecular Weight (g/mol): 60.056 MDL Number: MFCD00008022 InChI Key: XSQUKJJJFZCRTK-UHFFFAOYSA-N Synonym: carbamide,isourea,carbonyldiamide,ureophil,carbonyldiamine,carbamimidic acid,pseudourea,ureaphil,urevert,alphadrate PubChem CID: 1176 ChEBI: CHEBI:48376 IUPAC Name: urea SMILES: C(=O)(N)N
Urea, Ultra Pure, 5lb, MP Biomedicals
CAS: 57-13-6 Molecular Formula: CH4N2O Molecular Weight (g/mol): 60.056 InChI Key: XSQUKJJJFZCRTK-UHFFFAOYSA-N Synonym: carbamide,isourea,carbonyldiamide,ureophil,carbonyldiamine,carbamimidic acid,pseudourea,ureaphil,urevert,alphadrate PubChem CID: 1176 ChEBI: CHEBI:48376 IUPAC Name: urea SMILES: C(=O)(N)N
Urea, ≥98%, Molecular Biology Reagent Grade, MP Biomedicals™
CAS: 57-13-6 Molecular Formula: CH4N2O Molecular Weight (g/mol): 60.056 MDL Number: MFCD00008022 InChI Key: XSQUKJJJFZCRTK-UHFFFAOYSA-N Synonym: carbamide,isourea,carbonyldiamide,ureophil,carbonyldiamine,carbamimidic acid,pseudourea,ureaphil,urevert,alphadrate PubChem CID: 1176 ChEBI: CHEBI:48376 IUPAC Name: urea SMILES: C(=O)(N)N
Urea, MP Biomedicals
CAS: 57-13-6 Molecular Formula: CH4N2O Molecular Weight (g/mol): 60.056 MDL Number: MFCD00008022 InChI Key: XSQUKJJJFZCRTK-UHFFFAOYSA-N Synonym: carbamide,isourea,carbonyldiamide,ureophil,carbonyldiamine,carbamimidic acid,pseudourea,ureaphil,urevert,alphadrate PubChem CID: 1176 ChEBI: CHEBI:48376 IUPAC Name: urea SMILES: C(=O)(N)N
Urea, Reagent grade, MP Biomedicals™
CAS: 57-13-6 Molecular Formula: CH4N2O Molecular Weight (g/mol): 60.056 InChI Key: XSQUKJJJFZCRTK-UHFFFAOYSA-N Synonym: carbamide,isourea,carbonyldiamide,ureophil,carbonyldiamine,carbamimidic acid,pseudourea,ureaphil,urevert,alphadrate PubChem CID: 1176 ChEBI: CHEBI:48376 IUPAC Name: urea SMILES: C(=O)(N)N
Urea, 99.3+%
CAS: 57-13-6 Molecular Formula: CH4N2O Molecular Weight (g/mol): 60.056 MDL Number: MFCD00008022 InChI Key: XSQUKJJJFZCRTK-UHFFFAOYSA-N Synonym: carbamide,isourea,carbonyldiamide,ureophil,carbonyldiamine,carbamimidic acid,pseudourea,ureaphil,urevert,alphadrate PubChem CID: 1176 ChEBI: CHEBI:48376 IUPAC Name: urea SMILES: C(=O)(N)N
Urea, USP, JP, 98-102%, Spectrum™ Chemical
CAS: 57-13-6 Molecular Formula: CH4N2O Molecular Weight (g/mol): 60.06 InChI Key: XSQUKJJJFZCRTK-UHFFFAOYSA-N IUPAC Name: urea SMILES: NC(N)=O