accessibility menu, dialog, popup

UserName

Viewing profile as user:
ONHOHN

CAS RN 57-41-0

IUPAC Name: 5,5-diphenylimidazolidine-2,4-dione
Synonyms: silantin hindatal sanepil fenytoin dak phenytoin sodium prompt phenytoin sodium iphenylhydantoin didan tdc 250 fenitoin comital
Molecular Weight (g/mol): 252.27
Molecular Formula: C15H12N2O2
InChi Key: CXOFVDLJLONNDW-UHFFFAOYSA-N
SMILES: O=C1NC(=O)C(N1)(C1=CC=CC=C1)C1=CC=CC=C1
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (1)
  • (2)
  • (2)
Physical Form 
  • (2)
Solvent or Matrix 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (3)
  • (1)
  • (2)
  • (3)
  • (1)
  • (2)
  • (2)
  • (2)
  • (1)