CAS RN 57-43-2
CAS RN 57-43-2
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
CERILLIANT™ Amobarbital Solution, 1.0 mg/mL in Methanol, Sold by MilliporeSigma™ Supelco™
A Certified Spiking Solution™ for amobarbital testing methods by GC/MS or LC/MS for clinical toxicology, medication monitoring, forensic analysis, pain prescription monitoring, or urine drug testing applications.