accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 57-63-6

IUPAC Name: (1R,3aS,3bR,9bS,11aS)-1-ethynyl-11a-methyl-1H,2H,3H,3aH,3bH,4H,5H,9bH,10H,11H,11aH-cyclopenta[a]phenanthrene-1,7-diol
Synonyms: ethinyl estradiol ethinyl E2 estoral {[orion]} linoral ethinylestradiol jenapharm ethynyloestradiol 17 α-ethynyloestradiol ethinyl oestradiol effik anovlar 17a-ethynyloestradiol-17b
Molecular Weight (g/mol): 296.41
Molecular Formula: C20H24O2
InChi Key: BFPYWIDHMRZLRN-SLHNCBLASA-N
SMILES: C[C@]12CC[C@H]3[C@@H](CCC4=CC(O)=CC=C34)[C@@H]1CC[C@@]2(O)C#C
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (2)
  • (1)
Grade 
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)