accessibility menu, dialog, popup

UserName

Viewing profile as user:
ClClClClClClClCl

CAS RN 57-74-9

IUPAC Name: 1,3,4,7,8,9,10,10-octachlorotricyclo[5.2.1.0²,⁶]dec-8-ene
Synonyms: chlordane caswell no. 174 oktaterr clordano dichlorochlordene aspon-chlordane termi-ded chlordane , pur dowchlor topiclor 20
Molecular Weight (g/mol): 409.76
Molecular Formula: C10H6Cl8
InChi Key: BIWJNBZANLAXMG-UHFFFAOYNA-N
SMILES: ClC1CC2C(C1Cl)C1(Cl)C(Cl)=C(Cl)C2(Cl)C1(Cl)Cl
Solvent or Matrix 
  • (2)
  • (1)
Element 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results