accessibility menu, dialog, popup

UserName

Viewing profile as user:
OHONHNHOO

CAS RN 570-22-9

IUPAC Name: 1H-imidazole-4,5-dicarboxylic acid
Synonyms: 1H-imidazole-4,5-dicarboxylic acid
Molecular Weight (g/mol): 156.1
Molecular Formula: C5H4N2O4
InChi Key: ZEVWQFWTGHFIDH-UHFFFAOYSA-N
SMILES: OC(=O)C1=C(N=CN1)C(O)=O
Percent Purity 
  • (1)
  • (1)
  • (3)
  • (1)
Quantity 
  • (2)
  • (2)
  • (2)
Physical Form 
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (1)
  • (5)
  • (1)
  • (1)
  • (3)
  • (1)
  • (2)
  • (2)
  • (2)
  • (1)
  • (2)