accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 57021-49-5

IUPAC Name: 3,5-dimethyl-2-phenylthiophene
Synonyms: 3,5-dimethyl-2-phenylthiophene
Molecular Weight (g/mol): 188.29
Molecular Formula: C12H12S
InChi Key: RLISGGFOYMIIRQ-UHFFFAOYSA-N
SMILES: CC1=CC(C)=C(S1)C1=CC=CC=C1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results