accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 57042-08-7

IUPAC Name: 1,6-diethyl hex-3-enedioate
Synonyms: 1,6-diethyl hex-3-enedioate
Molecular Weight (g/mol): 200.23
Molecular Formula: C10H16O4
InChi Key: YIDKJPKYCLMJQW-UHFFFAOYSA-N
SMILES: CCOC(=O)CC=CCC(=O)OCC

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results