accessibility menu, dialog, popup

UserName

Viewing profile as user:
ONOOOH

CAS RN 57078-98-5

IUPAC Name: 4-(2,5-dioxo-2,5-dihydro-1H-pyrrol-1-yl)butanoic acid
Synonyms: N-maleoyl-4-aminobutyric acid
Molecular Weight (g/mol): 183.16
Molecular Formula: C8H9NO4
InChi Key: NCPQROHLJFARLL-UHFFFAOYSA-N
SMILES: OC(=O)CCCN1C(=O)C=CC1=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results