accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 57079-01-3

ONOOOH

CAS RN 57079-01-3

IUPAC Name: 11-(2,5-dioxo-2,5-dihydro-1H-pyrrol-1-yl)undecanoic acid
Synonyms: 11-(2,5-dioxopyrrol-1-yl)undecanoic acid
Molecular Weight (g/mol): 281.35
Molecular Formula: C15H23NO4
InChi Key: UVZTZBRGZXIBLZ-UHFFFAOYSA-N
SMILES: OC(=O)CCCCCCCCCCN1C(=O)C=CC1=O
Molecular Weight (g/mol) 
  • (1)
  • (3)
Quantity 
  • (1)
  • (2)
  • (1)
Percent Purity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results