accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OOFFFOOCH3(E/Z)

CAS RN 571-55-1

IUPAC Name: ethyl 2-(ethoxymethylidene)-4,4,4-trifluoro-3-oxobutanoate
Synonyms: ethyl 2-(ethoxymethylidene)-4,4,4-trifluoro-3-oxobutanoate
Molecular Weight (g/mol): 240.18
Molecular Formula: C9H11F3O4
InChi Key: XNGGOXOLHQANRB-UHFFFAOYSA-N
SMILES: CCOC=C(C(=O)OCC)C(=O)C(F)(F)F
Boiling Point 
  • (2)
  • (3)
Molecular Weight (g/mol) 
  • (4)
  • (1)
Percent Purity 
  • (2)
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (2)
  • (3)
  • (4)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)