accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 571-58-4

IUPAC Name: 1,4-dimethylnaphthalene
Synonyms: 1,4-dimethylnaphthalene epa pesticide chemical code 055802 1,4-dimethylnapthalene
Molecular Weight (g/mol): 156.23
Molecular Formula: C12H12
InChi Key: APQSQLNWAIULLK-UHFFFAOYSA-N
SMILES: CC1=C2C=CC=CC2=C(C)C=C1
Boiling Point 
  • (1)
  • (2)
Molecular Weight (g/mol) 
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)