accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CNBr

CAS RN 57102-97-3

IUPAC Name: 3-bromo-9-ethyl-9H-carbazole
Synonyms: 3-bromo-9-ethylcarbazole
Molecular Weight (g/mol): 274.16
Molecular Formula: C14H12BrN
InChi Key: PPHYYUDMDADQPF-UHFFFAOYSA-N
SMILES: CCN1C2=C(C=CC=C2)C2=C1C=CC(Br)=C2
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results