accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OHNOCH3

CAS RN 57103-01-2

IUPAC Name: 3,6-dimethoxy-9H-carbazole
Synonyms: 3,6-dimethoxy-9H-carbazole
Molecular Weight (g/mol): 227.26
Molecular Formula: C14H13NO2
InChi Key: YQKMWXHJSIEAEX-UHFFFAOYSA-N
SMILES: COC1=CC2=C(NC3=C2C=C(OC)C=C3)C=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results