accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3HNNNH2NNH2OCH3H3CH3C

CAS RN 57103-13-6

IUPAC Name: 5-[(8-methoxy-2,2,4-trimethyl-1,2,3,4-tetrahydroquinolin-6-yl)methyl]pyrimidine-2,4-diamine
Synonyms: 5-[(8-methoxy-2,2,4-trimethyl-3,4-dihydro-1H-quinolin-6-yl)methyl]pyrimidine-2,4-diamine
Molecular Weight (g/mol): 327.43
Molecular Formula: C18H25N5O
InChi Key: IPFLSUSUYBRVNV-UHFFFAOYNA-N
SMILES: COC1=C2NC(C)(C)CC(C)C2=CC(CC2=CN=C(N)N=C2N)=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results