accessibility menu, dialog, popup

UserName

Viewing profile as user:
NI

CAS RN 57103-15-8

IUPAC Name: 9-(4-iodophenyl)-9H-carbazole
Synonyms: 9-(4-iodophenyl)carbazole
Molecular Weight (g/mol): 369.21
Molecular Formula: C18H12IN
InChi Key: CWZDMKAGSIDFBR-UHFFFAOYSA-N
SMILES: IC1=CC=C(C=C1)N1C2=C(C=CC=C2)C2=C1C=CC=C2
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results