accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOOHO(E)

CAS RN 5711-41-1

IUPAC Name: (2E)-4-(4-methoxyphenyl)-4-oxobut-2-enoic acid
Synonyms: (2E)-4-(4-methoxyphenyl)-4-oxobut-2-enoic acid
Molecular Weight (g/mol): 206.2
Molecular Formula: C11H10O4
InChi Key: WORYXBDHTBWLLL-VOTSOKGWSA-N
SMILES: COC1=CC=C(C=C1)C(=O)\C=C\C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results