accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NClH3COCl

CAS RN 571159-05-2

IUPAC Name: 2-chloro-1-[1-(4-chlorophenyl)-2,5-dimethyl-1H-pyrrol-3-yl]ethan-1-one
Synonyms: 2-chloro-1-[1-(4-chlorophenyl)-2,5-dimethylpyrrol-3-yl]ethanone
Molecular Weight (g/mol): 282.16
Molecular Formula: C14H13Cl2NO
InChi Key: OYEPFHWXRWTABW-UHFFFAOYSA-N
SMILES: CC1=CC(C(=O)CCl)=C(C)N1C1=CC=C(Cl)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results