accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OON

CAS RN 5715-02-6

IUPAC Name: 2-cyanophenyl acetate
Synonyms: 2-cyanophenyl acetate
Molecular Weight (g/mol): 161.16
Molecular Formula: C9H7NO2
InChi Key: XXLKCUTUGWSJJO-UHFFFAOYSA-N
SMILES: CC(=O)OC1=CC=CC=C1C#N
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results