accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 5715-23-1

CH3OHH3C

CAS RN 5715-23-1

IUPAC Name: 3,4-dimethylcyclohexan-1-ol
Synonyms: 3,4-dimethyl-cyclohexanol
Molecular Weight (g/mol): 128.22
Molecular Formula: C8H16O
InChi Key: ZBAXJUPCYVIBSP-UHFFFAOYNA-N
SMILES: CC1CCC(O)CC1C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results